missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, Cell Culture Reagent

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

Supplier:  Thermo Scientific Chemicals J6190422

Quantity 100 g

 View more versions of this product

Catalog No. AAJ6190422


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63-68-3
149.21
FFEARJCKVFRZRR-BYPYZUCNSA-N
6137
CSCC[C@H](N)C(O)=O
C5H11NO2S
MFCD00063097 MFCD00801344
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
CHEBI:16643

Specifications

284°C (decomposition)
63-68-3
C5H11NO2S
1722294
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
FFEARJCKVFRZRR-BYPYZUCNSA-N
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
100 g
CHEBI:16643
Cell Culture Reagent
L-Methionine
White
MFCD00063097 MFCD00801344
14,5975
Soluble in water
CSCC[C@H](N)C(O)=O
149.21
6137
149.21
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 200-562-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions