missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, Cell Culture Reagent

Catalog No. p-7042262
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6190418 50 g
AAJ6190422 100 g
AAJ6190436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6190418 Supplier Thermo Scientific Chemicals Supplier No. J6190418

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 284°C (decomposition)
Color White
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
Quantity 50 g
Formula Weight 149.21
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Methionine
Show More Show Less
Hazard Category H500
EINECSNumber 200-562-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only