missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, 99+%, MP Biomedicals

$62.56

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN19470725
View Documents Promotion Details
N/A 25 g
Each for $62.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

63-68-3
149.21
FFEARJCKVFRZRR-BYPYZUCNSA-N
6137
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
C5H11NO2S
MFCD00063097 MFCD00801344
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
CHEBI:16643
CSCC[C@H](N)C(O)=O

Specifications

63-68-3
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
6137
149.2
Crystals
C5H11NO2S
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
CSCC[C@H](N)C(O)=O
149.21
CHEBI:16643
Cell Culture
L-Methionine, 99+%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS
Documents

Documents