missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, 99+%, MP Biomedicals

Catalog No. ICN19470725
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19470725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19470725 Supplier MP Biomedicals Inc Supplier No. 0219470725

missing translation for 'validMainframeMsg'

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Quantity 25 g
Formula Weight 149.2
Percent Purity 100%
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Methionine, 99+%
Recommended Storage Room temperature