missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Methioninamide hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 16120-92-6 |
|---|---|
| Molecular Formula | C5H13ClN2OS |
| Molecular Weight (g/mol) | 184.68 |
| MDL Number | MFCD00039084 |
| InChI Key | PWCBMJHINDTXGV-UHFFFAOYNA-N |
| Synonym | l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride |
| PubChem CID | 18531008 |
| IUPAC Name | (2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride |
| SMILES | Cl.CSCCC(N)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC426710010
|
Thermo Scientific Chemicals
426710010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC426710050
|
Thermo Scientific Chemicals
426710050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 16120-92-6 | |
| 184.68 | |
| PWCBMJHINDTXGV-UHFFFAOYNA-N | |
| 18531008 | |
| Cl.CSCCC(N)C(N)=O |
| C5H13ClN2OS | |
| MFCD00039084 | |
| l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride | |
| (2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride |
Specifications
| 16120-92-6 | |
| Glass bottle | |
| CH3SCH2CH2CH(CNH2)CONH2·HCl | |
| 1g | |
| PWCBMJHINDTXGV-UHFFFAOYNA-N | |
| (2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride | |
| 18531008 | |
| 98% |
| 97.5 to 102.5% (Argentometry) | |
| C5H13ClN2OS | |
| MFCD00039084 | |
| l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride | |
| Cl.CSCCC(N)C(N)=O | |
| 184.68 | |
| 184.69 | |
| L-Methioninamide hydrochloride |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only