missing translation for 'onlineSavingsMsg'
Learn More

L-Methioninamide hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 426710010

 View more versions of this product

Catalog No. AC426710010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L-Methioninamide hydrochloride
97.5 to 102.5% (Argentometry)
C5H13ClN2OS
MFCD00039084
l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride
Cl.CSCCC(N)C(N)=O
184.68
184.69
16120-92-6
Glass bottle
CH3SCH2CH2CH(CNH2)CONH2·HCl
1g
PWCBMJHINDTXGV-UHFFFAOYNA-N
(2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride
18531008
98%

Chemical Identifiers

16120-92-6
184.68
PWCBMJHINDTXGV-UHFFFAOYNA-N
18531008
Cl.CSCCC(N)C(N)=O
C5H13ClN2OS
MFCD00039084
l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride
(2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only