missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Menthol, 99.5%

CAS: 2216-51-5 | C10H20O | 156.27 g/mol

Supplier:  Thermo Scientific Chemicals 125401000

 View more versions of this product

Catalog No. AC125401000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Menthol
2216-51-5
Colorless to White
212.0°C
Authentic
Glass bottle
1.46
100 g
13,172; 16,203
15,5905
− 51.00
Solubility in water: insoluble. Other solubilities: freely soluble in acetic acid,soluble in alcohol,chloroform,ether,,petroleum ether
CC1CCC(C(C1)O)C(C)C
156.27
CHEBI:15409
99.5%
99.7%
41.0°C to 43.0°C
0.8900g/mL
94°C
99.4% min. (GC)
C10H20O
MFCD00062979
06,28
0.89
−51.00 (20.00°C 589nm,c=10,EtOH)
l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
16666
156.27
Crystals or Crystalline Needles

Chemical Identifiers

2216-51-5
156.27
NOOLISFMXDJSKH-KXUCPTDWSA-N
16666
(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
C10H20O
MFCD00062979
l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
CHEBI:15409
CC1CCC(C(C1)O)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 218-690-9

RTECSNumber : OT0700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only