missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Malic Acid 98.0+%, TCI America™
Supplier: TCI America M0022100G
Specifications
| L-(-)-Malic Acid | |
| 103°C | |
| C4H6O5 | |
| 100 g | |
| BJEPYKJPYRNKOW-REOHCLBHSA-N | |
| (2S)-2-hydroxybutanedioic acid | |
| 222656 | |
| 134.09 | |
| Crystalline Powder |
| 97-67-6 | |
| White | |
| MFCD00064213 | |
| l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid | |
| C(C(C(=O)O)O)C(=O)O | |
| 134.087 | |
| CHEBI:30797 | |
| ≥98.0% (T) |
Chemical Identifiers
| 97-67-6 | |
| 134.087 | |
| BJEPYKJPYRNKOW-REOHCLBHSA-N | |
| 222656 | |
| (2S)-2-hydroxybutanedioic acid |
| C4H6O5 | |
| MFCD00064213 | |
| l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid | |
| CHEBI:30797 | |
| C(C(C(=O)O)O)C(=O)O |
Safety and Handling
EINECSNumber : (2)-1442
RTECSNumber : ON7260000
TSCA : Yes