missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Malic Acid 98.0+%, TCI America™

Catalog No. p-7131932
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M002225G 25 g
M0022100G 100 g
M0022500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M002225G Supplier TCI America Supplier No. M002225G
Only null left

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 103°C
Color White
Quantity 25 g
Formula Weight 134.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Malic Acid
EINECSNumber (2)-1442
RTECSNumber ON7260000
TSCA Yes