missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-m-Tyrosine, 97+%
Description
An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in dimethyl sulfoxide, dilute acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
Soluble in dimethyl sulfoxide, dilute acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
Specifications
Specifications
| Melting Point | 260°C to 270°C |
| Solubility Information | Soluble in dimethyl sulfoxide,dilute acid. |
| Quantity | 1 g |
| Formula Weight | 181.19 |
| Percent Purity | ≥97% |
| Chemical Name or Material | L-m-Tyrosine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only