accessibility menu, dialog, popup

UserName

(S)ONH2OHHO

CAS RN 587-33-7

IUPAC Name: (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
Synonyms: meta-tyrosine
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: JZKXXXDKRQWDET-SVGMAFHSNA-N
SMILES: N[C@@H](CC1=CC=CC(O)=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)