missing translation for 'onlineSavingsMsg'
Learn More

L-m-Tyrosine, 97+%

Catalog No. AAH27224MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27224MD 250 mg
AAH2722403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27224MD Supplier Thermo Scientific Chemicals Supplier No. H27224MD

CAS: 587-33-7 | C9H11NO3 | 181.19 g/mol

An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, dilute acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).

Chemical Identifiers

CAS 587-33-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063059
InChI Key JZKXXXDKRQWDET-SVGMAFHSNA-N
Synonym l-m-tyrosine, 3-hydroxy-l-phenylalanine, meta-tyrosine, s-2-amino-3-3-hydroxyphenyl propanoic acid, 3-tyrosine, 3-hydroxyphenylalanine, m-tyrosine, l-m-tyr, unii-d5yf57v4qw, 3-m-hydroxyphenyl alanine
PubChem CID 6950578
ChEBI CHEBI:44303
IUPAC Name (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(O)=C1)C(O)=O

Specifications

Melting Point 260°C to 270°C
Solubility Information Soluble in dimethyl sulfoxide,dilute acid.
Quantity 250 mg
Formula Weight 181.19
Percent Purity ≥97%
Chemical Name or Material L-m-Tyrosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only