missing translation for 'onlineSavingsMsg'
Learn More

L-Lyxose, 99%

Catalog No. AAB2158303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2158303 1 g
AAB2158306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2158303 Supplier Thermo Scientific Chemicals Supplier No. B2158303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1949-78-6 | C5H10O5 | 150.13 g/mol

It acts as a reducing carbohydrate present in maple syrup. It is used in molecular modeling calculations in the study of drug binding and recognition in relation to aldose reductase. L-Lyxose is also utilized by Escherichia coli, encoding a kinase for L-Xylulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a reducing carbohydrate present in maple syrup. It is used in molecular modeling calculations in the study of drug binding and recognition in relation to aldose reductase. L-Lyxose is also utilized by Escherichia coli, encoding a kinase for L-Xylulose.

Solubility
Very soluble (586 g/L) (25°C) in water.

Notes
Hygroscopic. Store away from oxidizing agents, and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 1949-78-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064111
InChI Key PYMYPHUHKUWMLA-YUPRTTJUSA-N
Synonym l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci
PubChem CID 644176
ChEBI CHEBI:28480
IUPAC Name (2R,3R,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 108°C to 112°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Very soluble (586g/L) (25°C) in water.
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L-Lyxose
EINECSNumber 217-763-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only