missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Lysine 97.0+%, TCI America™
Supplier: TCI America L012925G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C6H14N2O2 | |
| MFCD00064433 | |
| l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
| CHEBI:18019 | |
| NCCCC[C@H](N)C(O)=O |
Specifications
| 212°C | |
| 56-87-1 | |
| C6H14N2O2 | |
| l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
| NCCCC[C@H](N)C(O)=O | |
| 25 g | |
| 5962 | |
| 146.19 | |
| Crystalline Powder |
| L-(+)-Lysine | |
| White-Yellow | |
| MFCD00064433 | |
| KDXKERNSBIXSRK-MLHKIVSYNA-N | |
| (2S)-2,6-diaminohexanoic acid | |
| 146.19 | |
| CHEBI:18019 | |
| ≥97.0% (T) |
Safety and Handling
EINECSNumber : (9)-1633
RTECSNumber : OL5540000
TSCA : Yes