missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine Monohydrochloride, FCC, 98.5-101.5%, Spectrum™ Chemical
L1143, 657-27-2, C6H14N2O2·HCl
Supplier: Spectrum Chemical Mfg Cor L1143500GM
| Quantity | 500 g |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical L-Lysine Monohydrochloride, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 657-27-2 | |
| 182.65 | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| Cl.NCCCC[C@H](N)C(O)=O |
| C6H15ClN2O2 | |
| MFCD00064564 MFCD00081870 | |
| (2S)-2,6-diaminohexanoic acid hydrochloride |
Specifications
| 657-27-2 | |
| 0.2% | |
| 1% | |
| Poly Bottle | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| (2S)-2,6-diaminohexanoic acid hydrochloride | |
| 500 g | |
| 98.5 to 101.5% |
| 100% | |
| 5 mg/kg | |
| C6H15ClN2O2 | |
| MFCD00064564 MFCD00081870 | |
| Cl.NCCCC[C@H](N)C(O)=O | |
| 182.65 | |
| +20.3 to +21.5° | |
| FCC |