missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrochloride, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603940
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603940 100 g Poly Bottle
18603941 500 g Poly Bottle
18603942 1 kg Poly Pail
18603943 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603940 Supplier Spectrum Chemical Mfg Cor Supplier No. L1143100GM

L1143, 657-27-2, C6H14N2O2·HCl

Spectrum™ Chemical L-Lysine Monohydrochloride, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Lead (Pb) 5 mg/kg
Loss on Drying 1%
Packaging Poly Bottle
Quantity 100 g
Optical Rotation +20.3 to +21.5°
Percent Purity 98.5 to 101.5%
Grade FCC