missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+%

Catalog No. AAA162490B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA162490B 1000 g
AAA1624918 50 g
AAA1624930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA162490B Supplier Thermo Scientific Chemicals Supplier No. A162490B
Only null left

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point ∼260°C (decomposition)
Beilstein 3563889
Merck Index 14,5636
Solubility Information It is soluble in water.
Quantity 1000 g
Formula Weight 182.65
Optical Rotation +20° (c=5 in 5N HCl)
Percent Purity ≥99%
Chemical Name or Material L-Lysine monohydrochloride
EINECSNumber 211-519-9
RTECSNumber OL5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only