missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrochloride, 99%

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

Supplier:  Thermo Scientific Chemicals 125222500

 View more versions of this product

Catalog No. AC125222500


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

263.0°C to 264.0°C
657-27-2
0.5% max. (TLC, 50μgspotting)
Authentic
0.4% max. (105°C, 3 hrs)
C6H15ClN2O2
H2N(CH2)4CH(NH2)CO2H·HCl
+ 21.00
15, 5697
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
BVHLGVCQOALMSV-JEDNCBNOSA-N
(2S)-2,6-diaminohexanoic acid hydrochloride
182.65
69568
182.65
Crystalline Powder or Crystals
L(+)-Lysine monohydrochloride
White
15ppm max.
30ppm max.
99+%
Plastic Bottle
MFCD00064564 MFCD00081870
300ppm max.
0.1% max.
Solubility in water: 65 g/100 mL (20°C).
Cl.NCCCC[C@H](N)C(O)=O
+21° (20°C c=8,6N HCl)
250 g
CHEBI:53633
≥99%

Chemical Identifiers

657-27-2
182.65
BVHLGVCQOALMSV-JEDNCBNOSA-N
69568
Cl.NCCCC[C@H](N)C(O)=O
C6H15ClN2O2
MFCD00064564 MFCD00081870
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
CHEBI:53633
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only