missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrochloride, 99%

Catalog No. AC125221000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125221000 100 g Plastic Bottle
AC125222500 250 g Plastic Bottle
AC125220010 1 kg Plastic Bottle
AC125220050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125221000 Supplier Thermo Scientific Chemicals Supplier No. 125221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 263.0°C to 264.0°C
Color White
Foreign Amino Acids 0.5% max. (TLC, 50μgspotting)
Heavy Metals (as Pb) 15ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99+%
Loss on Drying 0.4% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula H2N(CH2)4CH(NH2)CO2H·HCl
Specific Rotation + 21.00
Sulfate 300ppm max.
Merck Index 15, 5697
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 65 g/100 mL (20°C).
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
Specific Rotation Condition +21° (20°C c=8,6N HCl)
Quantity 100 g
Formula Weight 182.65
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Lysine monohydrochloride
Show More Show Less

RUO – Research Use Only