missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrochloride, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18611367
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611367 500 g Poly Pail
18611365 100 g Poly Pail
18611363 1 kg Poly Pail
18611364 5 kg Poly Pail
18611366 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611367 Supplier Spectrum Chemical Mfg Cor Supplier No. LY105500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 657-27-2 | 182.65 g/mol

Chemical Identifiers

CAS 657-27-2

Specifications

Arsenic (As) 1.5e-06
CAS Max % 1
Heavy Metals 1e-05
Iron (Fe) 3e-05
Loss on Drying 0.01
Packaging Poly Pail
Residue after Ignition 0.002
Specific Rotation +20.3° to +21.3°
Molecular Weight (g/mol) 182.65
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-Lysine Monohydrochloride
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.