missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99%

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

$148.12 - $148.12

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173730250
SDS View Documents
Thermo Scientific Chemicals
173730250
25 g Plastic Bottle
Each for $148.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39665-12-8
164.21
HZRUTVAFDWTKGD-XZNNNFJINA-N
16211825
C6H16N2O3
MFCD00151035
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
O.NCCCC[C@H](N)C(O)=O

Specifications

212.0°C to 214.0°C
39665-12-8
White to Yellow
10ppm max.
10ppm max.
0.5% max. (TLC)
Plastic Bottle
MFCD00151035
04, 435
95% min. (c=5, H2O, 430nm) 1cm cell
0.1% max.
10 to 11.5%
HZRUTVAFDWTKGD-XZNNNFJINA-N
(2S)-2,6-diaminohexanoic acid hydrate
164.21
16211825
99%
L(+)-Lysine monohydrate
200ppm max.
9 to 10.5 (2.5% soln. at 25°C)
300ppm max.
Authentic
99%
C6H16N2O3
H2N(CH2)4CH(NH2)CO2H·H2O
+ 26.25
200ppm max.
14, 5636
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
Solubility in water: 1500g/L (20°C). Other solubilities: insoluble in common neutral solvents
O.NCCCC[C@H](N)C(O)=O
+ 26.25 (20.00°C c=8,6N HCl)
25 g
164.21
Crystalline Powder or Crystals
Videos
SDS
Documents

Documents

RUO – Research Use Only