missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine monohydrate, 97%

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

Supplier:  Thermo Scientific Chemicals A1304022

 View more versions of this product

Catalog No. AAA1304022


Only null left

Description

Description

L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

Solubility
Soluble in water and hydrochloric acid.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

210°C (decomposition)
39665-12-8
MFCD00151035
14,5636
Soluble in water and hydrochloric acid.
O.NCCCC[C@H](N)C(O)=O
100 g
16211825
164.21 (146.19 Anhydrous)
97%
L-Lysine monohydrate
C6H16N2O3
1722531
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
HZRUTVAFDWTKGD-XZNNNFJINA-N
(2S)-2,6-diaminohexanoic acid;hydrate
164.21
+22.5° (c=2 in 6M HCl)
Air sensitive; Hygroscopic

Chemical Identifiers

39665-12-8
164.21
HZRUTVAFDWTKGD-XZNNNFJINA-N
16211825
C6H16N2O3
MFCD00151035
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
O.NCCCC[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-294-2

RTECSNumber : OL5540000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only