missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine Methyl Ester Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America L020225G
Specifications
| L-Lysine Methyl Ester Dihydrochloride | |
| 197°C | |
| C7H18Cl2N2O2 | |
| 25 g | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| dihydrogen methyl (2S)-2,6-diaminohexanoate dichloride | |
| 117778 | |
| ≥98.0% (N) |
| 26348-70-9 | |
| White | |
| MFCD00039067 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN | |
| 233.13 | |
| 233.13 | |
| Crystalline Powder |
Chemical Identifiers
| 26348-70-9 | |
| 233.13 | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| 117778 | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN |
| C7H18Cl2N2O2 | |
| MFCD00039067 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| dihydrogen methyl (2S)-2,6-diaminohexanoate dichloride |
Safety and Handling
TSCA : No