missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Methyl Ester Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7131866
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02025G 5 g
L020225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02025G Supplier TCI America Supplier No. L02025G
Only null left

Chemical Identifiers

CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Molecular Weight (g/mol) 233.13
MDL Number MFCD00039067
InChI Key SXZCBVCQHOJXDR-ILKKLZGPSA-N
Synonym l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
PubChem CID 117778
IUPAC Name dihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN

Specifications

Melting Point 197°C
Color White
Quantity 5 g
Formula Weight 233.13
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Lysine Methyl Ester Dihydrochloride