missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca L - Lysine Hydrochloride, USP
Description
L - Lysine Hydrochloride, USP is a white, odorless powder.
Alternative name(s): Lysine dihydrochloride
Specifications
Specifications
| Chemical Name or Material | L - Lysine Hydrochloride, USP |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 657-27-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 182.6495 |
| Quantity | 1 kg |
| Percent Purity | 98.5 to 101.5 % |
| Grade | USP |