missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine hydrochloride Solution, 100 mM Amino Acid in 0.1 M HCl, MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 10098-89-2 |
| Linear Formula | C6H14N2O2 · HCl |
| MDL Number | MFCD00064564 |
| Type | Analytical Standard |
| Quantity | 5 mL |
| Formula Weight | 182.65 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 mM amino acid in 0.1 M HCl |