missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Lysine, Hydrochloride, Calbiochem™,

Supplier:  MilliporeSigma™ 44001KG

 View more versions of this product

Catalog No. 44001KG


Only null left

Description

Description

Specifications

Specifications

657-27-2
C6H15ClN2O2
MFCD00064564 MFCD00081870
BVHLGVCQOALMSV-JEDNCBNOSA-N
(2S)-2,6-diaminohexanoic acid hydrochloride
182.65
CHEBI:53633
Solid
White
Glass Bottle
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
Cl.NCCCC[C@H](N)C(O)=O
1 kg
69568
≥98% (Titration (dry basis))

Chemical Identifiers

657-27-2
182.65
BVHLGVCQOALMSV-JEDNCBNOSA-N
69568
(2S)-2,6-diaminohexanoic acid hydrochloride
C6H15ClN2O2
MFCD00064564 MFCD00081870
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
CHEBI:53633
Cl.NCCCC[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions