missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Lysine, Hydrochloride, Calbiochem™,

Catalog No. 4400100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4400100GM 100 g
44001KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4400100GM Supplier MilliporeSigma™ Supplier No. 4400100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Format Glass Bottle

Specifications

Color White
Form Solid
Molecular Formula C6H15ClN2O2
MDL Number MFCD00064564 MFCD00081870
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES Cl.NCCCC[C@H](N)C(O)=O
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
Molecular Weight (g/mol) 182.65
PubChem CID 69568
ChEBI CHEBI:53633
Quantity 100 g
Percent Purity ≥98% (Titration (dry basis))
Chemical Name or Material L-Lysine Hydrochloride
Show More Show Less