missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine dihydrochloride, 99%
CAS: 657-26-1 | C6H16Cl2N2O2 | 219.11 g/mol
Supplier: Thermo Scientific Chemicals 413360250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 202.0°C | |
| 99% | |
| White to Yellow | |
| 0.5% max. (105°C, 3 hrs) | |
| C6H16Cl2N2O2 | |
| NH2(CH2)4CH(NH2)COOH·2HCl | |
| + 14.00 | |
| l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2 | |
| Cl.Cl.NCCCC[C@H](N)C(O)=O | |
| +14.00 (18.00°C c=2,water) | |
| 25 g | |
| 219.11 | |
| Crystalline Powder |
| L-Lysine dihydrochloride | |
| 657-26-1 | |
| Authentic | |
| 99% | |
| Glass Bottle | |
| MFCD00066389 | |
| 14, 5636 | |
| JBBURJFZIMRPCZ-XRIGFGBMSA-N | |
| (2S)-2,6-diaminohexanoic acid;dihydrochloride | |
| 219.11 | |
| 69567 | |
| 99% |
Chemical Identifiers
| 657-26-1 | |
| 219.11 | |
| JBBURJFZIMRPCZ-XRIGFGBMSA-N | |
| 69567 |
| C6H16Cl2N2O2 | |
| MFCD00066389 | |
| l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2 | |
| Cl.Cl.NCCCC[C@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 211-518-3
TSCA : TSCA
RUO – Research Use Only