missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine dihydrochloride, 99%

CAS: 657-26-1 | C6H16Cl2N2O2 | 219.11 g/mol

$107.88 - $107.88

Chemical Identifiers

CAS 657-26-1
Molecular Formula C6H16Cl2N2O2
Molecular Weight (g/mol) 219.11
MDL Number MFCD00066389
InChI Key JBBURJFZIMRPCZ-XRIGFGBMSA-N
Synonym l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
PubChem CID 69567
SMILES Cl.Cl.NCCCC[C@H](N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC413360250
SDS View Documents
Thermo Scientific Chemicals
413360250
25 g Glass Bottle
Each for $107.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

657-26-1
219.11
JBBURJFZIMRPCZ-XRIGFGBMSA-N
69567
C6H16Cl2N2O2
MFCD00066389
l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
Cl.Cl.NCCCC[C@H](N)C(O)=O

Specifications

202.0°C
White to Yellow
0.5% max. (105°C, 3 hrs)
C6H16Cl2N2O2
NH2(CH2)4CH(NH2)COOH·2HCl
+ 14.00
l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
Cl.Cl.NCCCC[C@H](N)C(O)=O
+14.00 (18.00°C c=2,water)
25 g
219.11
Crystalline Powder
657-26-1
Authentic
99%
Glass Bottle
MFCD00066389
14, 5636
JBBURJFZIMRPCZ-XRIGFGBMSA-N
(2S)-2,6-diaminohexanoic acid;dihydrochloride
219.11
69567
99%
L-Lysine dihydrochloride, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-518-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only