missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine acetate, 97%
CAS: 57282-49-2 | C8H18N2O4 | 206.24 g/mol
Supplier: Thermo Scientific Chemicals 451631000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Lysine acetate | |
| 97% | |
| MFCD00039069 | |
| RRNJROHIFSLGRA-JEDNCBNOSA-N | |
| acetic acid;(2S)-2,6-diaminohexanoic acid | |
| 206.24 | |
| 206.24 |
| 57282-49-2 | |
| C8H18N2O4 | |
| l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz | |
| CC(O)=O.NCCCC[C@H](N)C(O)=O | |
| 100 g | |
| 104152 | |
| 97% |
Chemical Identifiers
| 57282-49-2 | |
| 206.24 | |
| RRNJROHIFSLGRA-JEDNCBNOSA-N | |
| 104152 | |
| CC(O)=O.NCCCC[C@H](N)C(O)=O |
| C8H18N2O4 | |
| MFCD00039069 | |
| l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz | |
| acetic acid;(2S)-2,6-diaminohexanoic acid |
RUO – Research Use Only