accessibility menu, dialog, popup

UserName

OOHNH2(S)H2NOH3COH

CAS RN 57282-49-2

IUPAC Name: (2S)-2,6-diaminohexanoic acid; acetic acid
Synonyms: lysine acetate
Molecular Weight (g/mol): 206.24
Molecular Formula: C8H18N2O4
InChi Key: RRNJROHIFSLGRA-JEDNCBNOSA-N
SMILES: CC(O)=O.NCCCC[C@H](N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)