missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine, 98%

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol

Supplier:  Thermo Scientific Chemicals J6222522

Quantity 100 g

 View more versions of this product

Catalog No. AAJ6222522


Only null left

Description

Description

L-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.

Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvent.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

56-87-1
146.19
KDXKERNSBIXSRK-MLHKIVSYNA-N
5962
NCCCC[C@H](N)C(O)=O
C6H14N2O2
MFCD00064433
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
CHEBI:18019

Specifications

215°C
56-87-1
C6H14N2O2
1722531
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
KDXKERNSBIXSRK-MLHKIVSYNA-N
2,6-diaminohexanoic acid
146.19
CHEBI:18019
+26° (c=2 in 6M HCl)
Powder
L-Lysine
White
MFCD00064433
14,5636
Soluble in water; Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvent.
NCCCC[C@H](N)C(O)=O
100 g
5962
146.19
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-294-2

RTECSNumber : OL5540000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions