Learn More
Thermo Scientific™ L-Lysine-2HCl, 13C6 for SILAC
Description
Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.
Highlights:
Efficient – 100% label incorporation into proteins of living cells
Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
High-quality supplements – heavy amino acids with >99% isotope purity
Specifications
Specifications
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | Amino Acids |
| Content And Storage | Store at room temperature. |
| Label or Dye | Mass Isotope of Amino Acid |
| Shipping Condition | Room Temperature |
| Starting Material | Cells |
| Final Product Type | Protein |
| Detection Method | Mass Spectrometry |
| Workflow Step | Protein Labeling |
| Quantity | 50 mg |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.