missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Lysine-2HCl, 13C6, 15N2 for SILAC

Catalog No. p-7184498
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88209 50 mg
PI88432 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88209 Supplier Thermo Scientific™ Supplier No. 88209

Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.

Thermo Scientific Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits. This is a lyophilized preparation of L-Lysine-2HCl (13C6, 15N2) for SILAC experiments.

General features of heavy and light amino acids for SILAC labeling:

  • Efficient—100% label incorporation into proteins of living cells
  • Flexible—different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
  • Multiplex capabilities—several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
  • High-quality supplements—heavy amino acids with >99% isotope purity

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Thermo Scientific Heavy and Light Amino Acids for SILAC are used together with specialized cell culture media that are deficient in essential amino acids. Heavy and light L-lysine and L-arginine are the most common amino acids used for SILAC analysis of tryptic peptides. Up to three different experimental conditions can be readily analyzed with different isotopes of lysine and arginine. For lysine three-plex experiments, 4,4,5,5-D4 L-lysine and 13C6 15N2 L-lysine are used to generate peptides with 4- and 8-Da mass shifts, respectively, compared to peptides generated with light lysine. For arginine three-plex experiments, 13C6 L-arginine and 13C6 15N4 L-arginine are used to generate peptides with 6- and 10-Da mass shifts, respectively, compared to peptides generated with light arginine. L-leucine is another amino acid commonly used for SILAC labeling, because it is one of the most common amino acids found in protein sequences. Proline is a non-essential amino acid that is sometimes added to SILAC media to prevent the metabolic conversion of heavy arginine to heavy proline in mammalian cell lines with high arginine dehydrogenase activity.

Related Products

  • L-Arginine-HCl for SILAC (Cat. No. 89989)
  • L-Proline for SILAC (Cat. No. 88211)
  • L-Leucine for SILAC (Cat. No. 88428)
  • L-Lysine-2HCl for SILAC (Cat. No. 89987)

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Amino Acids
Content And Storage Store at room temperature.
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
Starting Material Cells
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 50 mg
Form Powder
Show More Show Less
Do you offer any heavy amino acids other than the ones in the SILAC Protein Quantitation Kits?

We offer additional isotopes of lysine, including 4,4,5,5 D4 L-Lysine-2HCl (Cat. No. 88437) and 13C6 15N2 L-Lysine-2HCl (Cat. No. 88209), which can be used for multiplexed experiments. 13C6 L-Arginine-HCl (Cat. No. 88210) and 13C6 15N2 L-Arginine-HCl (Cat. No. 89990) are also available to enhance peptide isotope label coverage of tryptic peptides. In addition to lysine and arginine, 13C6 L-Leucine (Cat. No. 88435) can also be used with our powdered SILAC DMEM (Cat. No. 88425) and RPMI-1640 media (Cat. No. 88426). Please check the product page for a current listing.

For Research Use Only. Not for use in diagnostic procedures.