missing translation for 'onlineSavingsMsg'
Learn More

L-Lysinamide dihydrochloride, 95%

Catalog No. p-7056675
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65741MD 250 mg
AAH6574103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH65741MD Supplier Thermo Scientific Chemicals Supplier No. H65741MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51127-08-3 | C6H17Cl2N3O | 218.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51127-08-3
Molecular Formula C6H17Cl2N3O
Molecular Weight (g/mol) 218.12
MDL Number MFCD00058287
InChI Key AIYVJLPYZQDCKV-XRIGFGBMSA-N
Synonym s-2,6-diaminohexanamide dihydrochloride, h-lys-nh2 2hcl, l-lysinamide dihydrochloride, h-lys-nh 2 hcl, lysine amide dihydrochloride, c6h15n3o.2hcl, h-lys-nh2 . 2 hcl
PubChem CID 20760365
IUPAC Name (2S)-2,6-diaminohexanamide dihydrochloride
SMILES Cl.Cl.NCCCC[C@H](N)C(N)=O

Specifications

Quantity 250 mg
Formula Weight 218.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material L-Lysinamide dihydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only