accessibility menu, dialog, popup

UserName

CAS RN 51127-08-3

IUPAC Name: (2S)-2,6-diaminohexanamide dihydrochloride
Synonyms: L-lysinamide dihydrochloride
Molecular Weight (g/mol): 218.12
Molecular Formula: C6H17Cl2N3O
InChi Key: AIYVJLPYZQDCKV-XRIGFGBMSA-N
SMILES: Cl.Cl.NCCCC[C@H](N)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)