missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucyl-L-tyrosine 98.0+%, TCI America™
$77.00 - $540.97
Chemical Identifiers
| CAS | 968-21-8 |
|---|---|
| Molecular Formula | C15H22N2O4 |
| Molecular Weight (g/mol) | 294.351 |
| MDL Number | MFCD00020195 |
| InChI Key | LHSGPCFBGJHPCY-STQMWFEESA-N |
| Synonym | l-leucyl-l-tyrosine, s-2-s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, chembl56099, leucyl-tyrosine, 2s-2-2s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid, n-l-leucyl-l-tyrosine, tyrosine, leucyl, h-leu-tyr-oh, l-leu-l-tyr |
| PubChem CID | 70410 |
| ChEBI | CHEBI:73591 |
| IUPAC Name | (2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L0040100MG
|
TCI America
L0040100MG |
100 mg |
Each for $77.00
|
|
|||||
|
L00401G
|
TCI America
L00401G |
1 g |
Each for $540.97
|
|
|||||
Chemical Identifiers
| 968-21-8 | |
| 294.351 | |
| LHSGPCFBGJHPCY-STQMWFEESA-N | |
| 70410 | |
| (2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid |
| C15H22N2O4 | |
| MFCD00020195 | |
| l-leucyl-l-tyrosine, s-2-s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, chembl56099, leucyl-tyrosine, 2s-2-2s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid, n-l-leucyl-l-tyrosine, tyrosine, leucyl, h-leu-tyr-oh, l-leu-l-tyr | |
| CHEBI:73591 | |
| CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N |
Specifications
| 968-21-8 | |
| C15H22N2O4 | |
| 100 mg | |
| LHSGPCFBGJHPCY-STQMWFEESA-N | |
| (2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid | |
| 70410 | |
| 294.35 | |
| Crystalline Powder |
| White | |
| MFCD00020195 | |
| l-leucyl-l-tyrosine, s-2-s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, chembl56099, leucyl-tyrosine, 2s-2-2s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid, n-l-leucyl-l-tyrosine, tyrosine, leucyl, h-leu-tyr-oh, l-leu-l-tyr | |
| CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N | |
| 294.351 | |
| CHEBI:73591 | |
| ≥98.0% (T) | |
| L-Leucyl-L-tyrosine |
Safety and Handling
TSCA : No