missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
$42.87 - $130.48
Chemical Identifiers
| CAS | 2748-02-9 |
|---|---|
| Molecular Formula | C10H22ClNO2 |
| Molecular Weight (g/mol) | 223.74 |
| MDL Number | MFCD00038906 |
| InChI Key | RFUWRXIYTQGFGA-QRPNPIFTSA-N |
| Synonym | h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl |
| PubChem CID | 12451443 |
| IUPAC Name | tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride |
| SMILES | Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C |
Chemical Identifiers
| 2748-02-9 | |
| 223.74 | |
| RFUWRXIYTQGFGA-QRPNPIFTSA-N | |
| 12451443 | |
| Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C |
| C10H22ClNO2 | |
| MFCD00038906 | |
| h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl | |
| tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride |
Specifications
| 2748-02-9 | |
| White-Yellow | |
| MFCD00038906 | |
| h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl | |
| Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C | |
| 223.74 | |
| 223.74 | |
| Crystalline Powder |
| 167°C | |
| C10H22ClNO2 | |
| 1 g | |
| RFUWRXIYTQGFGA-QRPNPIFTSA-N | |
| tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride | |
| 12451443 | |
| ≥98.0% (T) | |
| L-Leucine tert-Butyl Ester Hydrochloride |
Safety and Handling
TSCA : No