CAS RN 2748-02-9
IUPAC Name:
tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride
Synonyms:
tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride
Molecular Weight (g/mol):
223.74
Molecular Formula:
C10H22ClNO2
InChi Key:
RFUWRXIYTQGFGA-QRPNPIFTSA-N
SMILES:
Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C
1
–
2
of
2
results
1
L-Leucine tert-butyl ester hydrochloride, 98%
CAS: 2748-02-9 MDL Number: MFCD00038906 ChEBI: CHEBI:78502
2
L-Leucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
CAS: 2748-02-9 Molecular Formula: C10H22ClNO2 Molecular Weight (g/mol): 223.74 MDL Number: MFCD00038906 InChI Key: RFUWRXIYTQGFGA-QRPNPIFTSA-N Synonym: h-leu-otbu.hcl,l-leucine tert-butyl ester hydrochloride,h-leu-otbu hcl,l-leucine t-butyl ether hcl salt,l-leucine t-butyl ester hydrochloride,l-leucine tert-butyl ester hcl,tert-butyl l-leucinate hydrochloride,s-tert-butyl 2-amino-4-methylpentanoate hydrochloride,tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride,h-leu-obut hcl PubChem CID: 12451443 IUPAC Name: tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride SMILES: Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C