missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. L01971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01971G 1 g
L01975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01971G Supplier TCI America Supplier No. L01971G

Chemical Identifiers

CAS 2748-02-9
Molecular Formula C10H22ClNO2
Molecular Weight (g/mol) 223.74
MDL Number MFCD00038906
InChI Key RFUWRXIYTQGFGA-QRPNPIFTSA-N
Synonym h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl
PubChem CID 12451443
IUPAC Name tert-butyl (2S)-2-amino-4-methylpentanoate hydrochloride
SMILES Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucine tert-Butyl Ester Hydrochloride