missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine tert-butyl ester hydrochloride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Molecular Formula | C10H22ClNO2 |
| Beilstein | 3697141 |
| Synonym | h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl |
| InChI Key | RFUWRXIYTQGFGA-QRPNPIFTSA-N |
| SMILES | Cl.CC(C)C[C@H](N)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl (2S)-2-amino-4-methylpentanoate;hydrochloride |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 223.74 |
| PubChem CID | 12451443 |
| Formula Weight | 223.74 |
| Show More |
Safety and Handling
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only