missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine Ethyl Ester Hydrochloride 99.0+%, TCI America™
$115.51 - $115.51
Chemical Identifiers
| CAS | 2743-40-0 |
|---|---|
| Molecular Formula | C8H17NO2 |
| Molecular Weight (g/mol) | 159.23 |
| MDL Number | MFCD00034879 |
| InChI Key | QIGLJVBIRIXQRN-ZETCQYMHSA-N |
| Synonym | l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl |
| PubChem CID | 11658447 |
| IUPAC Name | ethyl (2S)-2-amino-4-methylpentanoate |
| SMILES | CCOC(=O)[C@@H](N)CC(C)C |
Chemical Identifiers
| 2743-40-0 | |
| 159.23 | |
| QIGLJVBIRIXQRN-ZETCQYMHSA-N | |
| 11658447 | |
| CCOC(=O)[C@@H](N)CC(C)C |
| C8H17NO2 | |
| MFCD00034879 | |
| l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl | |
| ethyl (2S)-2-amino-4-methylpentanoate |
Specifications
| 2743-40-0 | |
| White | |
| MFCD00034879 | |
| l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl | |
| CCOC(=O)[C@@H](N)CC(C)C | |
| 159.23 | |
| 195.69 | |
| Crystalline Powder |
| 136°C | |
| C8H17NO2 | |
| 25 g | |
| QIGLJVBIRIXQRN-ZETCQYMHSA-N | |
| ethyl (2S)-2-amino-4-methylpentanoate | |
| 11658447 | |
| ≥99.0% (T) | |
| L-Leucine Ethyl Ester Hydrochloride |
Safety and Handling
TSCA : No