missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine-2-naphthylamide Hydrochloride (2-Naphthylamine free) 98.0+%, TCI America™

Catalog No. p-7131785
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00371G 1 g
L00375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00371G Supplier TCI America Supplier No. L00371G

Chemical Identifiers

CAS 893-36-7
Molecular Formula C16H21ClN2O
Molecular Weight (g/mol) 292.807
MDL Number MFCD00034972
InChI Key HTHKKWZMEVJWQF-RSAXXLAASA-N
PubChem CID 16219549
IUPAC Name (2S)-2-amino-4-methyl-N-naphthalen-2-ylpentanamide;hydrochloride
SMILES CC(C)CC(C(=O)NC1=CC2=CC=CC=C2C=C1)N.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 292.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucine-2-naphthylamide Hydrochloride (2-Naphthylamine free)