accessibility menu, dialog, popup

UserName

CAS RN 893-36-7

HClH3CCH3NH2OHN

CAS RN 893-36-7

IUPAC Name: hydrogen 2-amino-4-methyl-N-(naphthalen-2-yl)pentanamide chloride
Synonyms: hydrogen 2-amino-4-methyl-N-(naphthalen-2-yl)pentanamide chloride
Molecular Weight (g/mol): 292.81
Molecular Formula: C16H21ClN2O
InChi Key: HTHKKWZMEVJWQF-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)CC(N)C(=O)NC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results