missing translation for 'onlineSavingsMsg'
Learn More

L-Leucic Acid 99.0+%, TCI America™

Catalog No. L00261G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00261G 1 g
L00265G 5 g
L002625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L00261G Supplier TCI America Supplier No. L00261G

Chemical Identifiers

CAS 13748-90-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00064214
InChI Key LVRFTAZAXQPQHI-YFKPBYRVSA-N
Synonym l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
PubChem CID 83697
ChEBI CHEBI:44510
IUPAC Name (2S)-2-hydroxy-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)O

Specifications

Melting Point 81°C
Color White
Quantity 1 g
Formula Weight 132.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucic Acid
EINECSNumber (2)-1340
TSCA Yes