missing translation for 'onlineSavingsMsg'
Learn More

L-Kynurenine

Catalog No. AAJ60199MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60199MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60199MF Supplier Thermo Scientific Chemicals Supplier No. J60199MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2922-83-0 | C10H12N2O3 | 208.22 g/mol

Intermediate in the breakdown of tryptophan

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2922-83-0
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.22
MDL Number MFCD00069912
InChI Key YGPSJZOEDVAXAB-QMMMGPOBSA-N
Synonym l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid
PubChem CID 161166
ChEBI CHEBI:16946
IUPAC Name (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
SMILES N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 208.21
Physical Form Crystalline
Chemical Name or Material L-Kynurenine
RTECSNumber CY9049700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only