missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Kynurenine Hydrate 98.0+%, TCI America™
Supplier: TCI America K0016100MG
Specifications
| L-Kynurenine Hydrate | |
| Yellow | |
| MFCD00069912 | |
| l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid | |
| N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O | |
| 208.22 | |
| CHEBI:16946 | |
| ≥98.0% (HPLC,T) |
| 2922-83-0 | |
| C10H12N2O3 | |
| 100 mg | |
| YGPSJZOEDVAXAB-QMMMGPOBSA-N | |
| (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid | |
| 161166 | |
| 208.22 | |
| Crystalline Powder |
Chemical Identifiers
| 2922-83-0 | |
| 208.22 | |
| YGPSJZOEDVAXAB-QMMMGPOBSA-N | |
| 161166 | |
| (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid |
| C10H12N2O3 | |
| MFCD00069912 | |
| l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid | |
| CHEBI:16946 | |
| N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O |
Safety and Handling
RTECSNumber : CY9049700
TSCA : No