missing translation for 'onlineSavingsMsg'
Learn More

L-Isoserine 98.0+%, TCI America™

Catalog No. p-7131618
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08281G 1 g
I08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08281G Supplier TCI America Supplier No. I08281G

Chemical Identifiers

CAS 632-13-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD03428326
InChI Key BMYNFMYTOJXKLE-REOHCLBHSA-N
Synonym l-isoserine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine, s-2-hydroxy-b-alanine
PubChem CID 6999797
IUPAC Name (2S)-3-azaniumyl-2-hydroxypropanoate
SMILES [NH3+]C[C@H](O)C([O-])=O

Specifications

Melting Point 197°C
Color White
Quantity 1 g
Formula Weight 105.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Isoserine
EINECSNumber (2)-4116
TSCA No