accessibility menu, dialog, popup

UserName

NH3(S)OHOO

CAS RN 632-13-3

IUPAC Name: (2S)-3-azaniumyl-2-hydroxypropanoate
Synonyms: (2S)-3-ammonio-2-hydroxypropanoate
Molecular Weight (g/mol): 105.09
Molecular Formula: C3H7NO3
InChi Key: BMYNFMYTOJXKLE-REOHCLBHSA-N
SMILES: [NH3+]C[C@H](O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)